EMAIL THIS PAGE TO A FRIEND
V2000 Sigma-Aldrich

Val-Trp

  • CAS Number 24587-37-9

  • Empirical Formula (Hill Notation) C16H21N3O3

  • Molecular Weight 303.36

  •  MDL number MFCD00037947

Purchase

Properties

storage temp.   −20°C
SMILES string   CC(C)[C@H](N)C(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(O)=O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?