• USA Home
  • V2627 - L-Valine benzyl ester p-toluenesulfonate salt

EMAIL THIS PAGE TO A FRIEND
V2627 Sigma-Aldrich

L-Valine benzyl ester p-toluenesulfonate salt

  • CAS Number 16652-76-9

  • Linear Formula (CH3)2CHCH(NH2)COOCH2C6H5 · CH3C6H4SO3H

  • Molecular Weight 379.47

  •  Beilstein/REAXYS Number 3643436

  •  EC Number 240-702-6

  •  MDL number MFCD00038908

  •  PubChem Substance ID 24900735

Purchase

Properties

Related Categories Amino Acids, Amino Acids & Derivatives, Biochemicals and Reagents, Chemical Biology, Chemical Synthesis,
form   powder
color   white
mp   160-160 °C
storage temp.   −20°C
SMILES string   Cc1ccc(cc1)S(O)(=O)=O.CC(C)[C@H](N)C(=O)OCc2ccccc2
InChI   1S/C12H17NO2.C7H8O3S/c1-9(2)11(13)12(14)15-8-10-6-4-3-5-7-10;1-6-2-4-7(5-3-6)11(8,9)10/h3-7,9,11H,8,13H2,1-2H3;2-5H,1H3,(H,8,9,10)/t11-;/m0./s1
InChI key   QWUQVUDPBXFOKF-MERQFXBCSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?