EMAIL THIS PAGE TO A FRIEND
V6264 Sigma-Aldrich

Venturicidin A

from Streptomyces sp., ≥95% (HPLC), lyophilized powder

Synonym: Aabomycin A1

Purchase

Properties

biological source   Streptomyces sp.
assay   ≥95% (HPLC)
form   lyophilized powder
solubility   DMF: soluble
  DMSO: soluble
  ethanol: soluble
  methanol: soluble
Mode of action   enzyme | inhibits
antibiotic activity spectrum   fungi
storage temp.   2-8°C

Description

Biochem/physiol Actions

Antifungal antibiotic; uncoupler of mitchondrial oxidative phosphorylation, F0F1-ATPase (ATP synthase) inhibitor.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?