• USA Home
  • V8626 - Val-Pro-Pro-Pro-Val-Pro-Pro-Arg-Arg-Arg

EMAIL THIS PAGE TO A FRIEND
V8626 Sigma-Aldrich

Val-Pro-Pro-Pro-Val-Pro-Pro-Arg-Arg-Arg

≥97% (CE)

Synonym: Ras inhibitory peptide

  • CAS Number 159088-48-9

  • Empirical Formula (Hill Notation) C53H91N19O11

  • Molecular Weight 1170.41

  •  MDL number MFCD00237248

Purchase

Properties

assay   ≥97% (CE)
composition   Peptide content, ~60%
storage temp.   −20°C
SMILES string   CC(C)[C@H](N)C(=O)N1CCC[C@H]1C(=O)N2CCC[C@H]2C(=O)N3CCC[C@H]3C(=O)N[C@@H](C(C)C)C(=O)N4CCC[C@H]4C(=O)N5CCC[C@H]5C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

Description

Biochem/physiol Actions

Prevents binding of the Ras exchange factor hSos1 to the human adapter protein Grb2.

Other Notes

Fragment 1149-1158 of the guanine nucleotide-releasing factor hSos1.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?