EMAIL THIS PAGE TO A FRIEND
V9006 Sigma-Aldrich

Val-Thr-Lys-Gly

≥97% (HPLC)

  • CAS Number 133605-54-6

  • Empirical Formula (Hill Notation) C17H33N5O6

  • Molecular Weight 403.47

  •  MDL number MFCD00133918

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C
SMILES string   CC(C)C(N)C(=O)NC(C(C)O)C(=O)NC(CCCCN)C(=O)NCC(O)=O
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?