EMAIL THIS PAGE TO A FRIEND
W4013 Sigma-Aldrich

WP900 hydrochloride

≥95% (HPLC), solid

Synonym: (−)-Daunorubicin

  • Empirical Formula (Hill Notation) C27H30NO10Cl

  • Molecular Weight 563.98

  •  MDL number MFCD04974507

  •  PubChem Substance ID 24724666

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, DNA-RNA Transcription Regulators, Gene Regulation and Expression
assay   ≥95% (HPLC)
form   solid
storage temp.   −20°C
SMILES string   Cl[H].COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O[C@H]5C[C@H](N)[C@H](O)[C@H](C)O5)c4c(O)c3C(=O)c12)C(C)=O
InChI   1S/C27H29NO10.ClH/c1-10-22(30)14(28)7-17(37-10)38-16-9-27(35,11(2)29)8-13-19(16)26(34)21-20(24(13)32)23(31)12-5-4-6-15(36-3)18(12)25(21)33;/h4-6,10,14,16-17,22,30,32,34-35H,7-9,28H2,1-3H3;1H/t10-,14-,16-,17-,22+,27-;/m0./s1
InChI key   GUGHGUXZJWAIAS-QQYBVWGSSA-N
Gene Information   human ... TOP2A(7153)

Description

Biochem/physiol Actions

Synthetic left-handed enantiomer of (+)-daunorubicin, anti-tumor antibiotic, binds to right-handed B-form DNA under low salt conditions.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?