EMAIL THIS PAGE TO A FRIEND
X2126 Sigma-Aldrich

DL-Xylose

≥99%

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, Monosaccharide More...
Quality Level   200
assay   ≥99%
form   powder
application(s)   HPLC: suitable
SMILES string   O[C@@H]1COC(O)[C@H](O)[C@H]1O
InChI   1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4+,5+/m0/s1
InChI key   PYMYPHUHKUWMLA-VPENINKCSA-N

Description

Biochem/physiol Actions

Xylose is a aldopentose sugar first isolated from wood.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?