• USA Home
  • Z0537 - trans-Zeatin glucoside

EMAIL THIS PAGE TO A FRIEND
Z0537 Sigma-Aldrich

trans-Zeatin glucoside

~95%

Synonym: 9-(β-D-Glucopyranosyl)-trans-zeatin

  • CAS Number 51255-96-0

  • Empirical Formula (Hill Notation) C16H23N5O6

  • Molecular Weight 381.38

  •  MDL number MFCD00079650

  •  PubChem Substance ID 24902166

Purchase

Properties

assay   ~95%
storage temp.   −20°C
SMILES string   [H]\C(CNc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)=C(\C)CO
InChI   1S/C16H23N5O6/c1-8(4-22)2-3-17-14-10-15(19-6-18-14)21(7-20-10)16-13(26)12(25)11(24)9(5-23)27-16/h2,6-7,9,11-13,16,22-26H,3-5H2,1H3,(H,17,18,19)/b8-2+/t9-,11-,12+,13-,16-/m1/s1
InChI key   VYRAJOITMBSQSE-HNVSNYHQSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?