EMAIL THIS PAGE TO A FRIEND
Z1252 Sigma-Aldrich

Zofenopril calcium

>98% (HPLC), powder

Synonym: (4S)-1-[(2S)-3-(benzoylthio)-2-methyl-1-oxopropyl]-4-(phenylthio)-L-proline calcium salt

  • CAS Number 81938-43-4

  • Empirical Formula (Hill Notation) C22H22NO4S2 · 0.5Ca

  • Molecular Weight 448.58

  •  MDL number MFCD11858294

  •  PubChem Substance ID 329831603

Purchase

Properties

Related Categories Angiotensin Metabolism, Approved Therapeutics/Drug Candidates, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Bristol-Myers Squibb,
Quality Level   100
assay   >98% (HPLC)
form   powder
color   white to off-white
solubility   DMSO: >5 mg/mL
originator   Bristol-Myers Squibb
storage temp.   2-8°C
SMILES string   [Ca++].C[C@H](CSC(=O)c1ccccc1)C(=O)N2C[C@H](C[C@H]2C([O-])=O)Sc3ccccc3.C[C@H](CSC(=O)c4ccccc4)C(=O)N5C[C@H](C[C@H]5C([O-])=O)Sc6ccccc6
InChI   1S/2C22H23NO4S2.Ca/c2*1-15(14-28-22(27)16-8-4-2-5-9-16)20(24)23-13-18(12-19(23)21(25)26)29-17-10-6-3-7-11-17;/h2*2-11,15,18-19H,12-14H2,1H3,(H,25,26);/q;;+2/p-2/t2*15-,18+,19+;/m11./s1
InChI key   NSYUKKYYVFVMST-LETVYOFWSA-L

Description

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

Zofenopril is a long-lasting, lipophilic ACE inhibitor. Zofenopril is also an inhibitor of PEPT2, the predominant peptide transporter in kidney and choroid plexus.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

This compound is featured on the Angiotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Angiotensin Receptors

Different receptors/binding sites have been identified for the biologically active angiotensin (Ang) peptides, i.e. Ang II (1-8), Ang III (2-8), Ang IV (3-8) and Ang (1-7), based on the availability ...
Keywords: Atomic absorption spectroscopy, Cardiovascular, Catalysis, Cell proliferation, Gene expression, Ligands, Metabolites, Transduction

Antihypertensive Agents

Agents that inhibit angiontensin-converting enzymes (ACE) and angiotensin II formation are essential to cardiovascular medicine.1 These inhibitors are used not only to treat essential hypertension an...
Sami Barghshoon
BioFiles v7 n5, 2012, 5–20
Keywords: AGE, Antihypertensives, Cardiovascular, Clinical, Diuretics, PAGE, Reductions

Discover Bioactive Small Molecules for ADME/Tox

A significant number of drugs that fail in clinical trials have been associated with safety issues, including unexpected drug-drug interactions (DDI) or lack of efficacy due to poor pharmacokinetics....
Keywords: Absorption, Bioactive small molecules, Clinical, Metabolism

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?