EMAIL THIS PAGE TO A FRIEND
A0533 Sigma

L-Amphetamine

Synonym: (−)-α-Methylphenethylamine, Levamphetamine

Purchase

Properties

drug control   USDEA Schedule II; Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

Description

Biochem/physiol Actions

Induces release of catecholamines, especially dopamine, and blocks catecholamine reuptake

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGII
WGK Germany 
3
RTECS 
SH9050000

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Amphetamine Enantiomers (N-Trifluoroacetyl Derivative) on Astec® CHIRALDEX® G-PN

From our library of Articles, Sigma-Aldrich presents GC Analysis of Amphetamine Enantiomers (N-Trifluoroacetyl Derivative) on Astec® CHIRALDEX® G-PN
Keywords: Chromatography, Clinical, Flame ionization detector, Forensic, Gas chromatography, Pharmaceutical, Purification

GC Analysis of Amphetamine and Methamphetamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-PN

From our library of Articles, Sigma-Aldrich presents GC Analysis of Amphetamine and Methamphetamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-PN
Keywords: Chromatography, Clinical, Flame ionization detector, Forensic, Gas chromatography, Pharmaceutical, Purification

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?