EMAIL THIS PAGE TO A FRIEND
A0634 Sigma-Aldrich

DL-Ala-DL-Val

  • CAS Number 1999-46-8

  • Empirical Formula (Hill Notation) C8H16N2O3

  • Molecular Weight 188.22

  •  MDL number MFCD00038163

Purchase

Properties

storage temp.   −20°C
SMILES string   CC(C)[C@H](NC(=O)[C@H](C)N)C(O)=O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?