• USA Home
  • A1120AO-HH - DMT-dA(tac) Phosphoramidite

EMAIL THIS PAGE TO A FRIEND
A1120AO-HH Sigma-Aldrich

DMT-dA(tac) Phosphoramidite

configured for ÄKTA® and OligoPilot®

Synonym: DMT-dA(tac) amidite

Purchase

Properties

Related Categories ABI 3900, ABI 39X, Chemical Synthesis, DNA Phosphoramidites, Phosphoramidites and Reagents for Oligonucleotide Synthesis (Proligo Reagents),
InChI Key   INUFZOANRLMUCW-SDNJGBRXSA-N
type   for DNA synthesis
product line   Proligo Reagents
assay   ≥98% (31P-NMR)
  ≥98.0% (reversed phase HPLC)
packaging   pkg of 1 × 10 g (A112010-01)
application(s)   oligo synthesis: suitable
λ   conforms (UV/VIS Identity)
compatibility   configured for ÄKTA® and OligoPilot®
nucleoside profile   base: adenosine
base protecting group: TAC
5' protecting group: DMT
deprotection: fast
storage temp.   −20°C

Description

Legal Information

OligoPilot is a registered trademark of GE Healthcare

ÄKTA is a registered trademark of GE Healthcare

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?