EMAIL THIS PAGE TO A FRIEND
A1257 Sigma-Aldrich

7-Azaindole

≥99%

Synonym: 1H-Pyrrolo(2,3-b)pyridine

Purchase

Properties

assay   ≥99%
mp   105-107 °C (lit.)
SMILES string   c1cnc2[nH]ccc2c1
InChI   1S/C7H6N2/c1-2-6-3-5-9-7(6)8-4-1/h1-5H,(H,8,9)
InChI key   MVXVYAKCVDQRLW-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
UY8710000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

A95502 7-Azaindole, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?