• USA Home
  • A1291 - L-Aspartic acid amide

EMAIL THIS PAGE TO A FRIEND
A1291 Sigma-Aldrich

L-Aspartic acid amide

Synonym: Isoasparagine

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?