• USA Home
  • A145 - 1-Allyl-3,7-dimethyl-8-sulfophenylxanthine

EMAIL THIS PAGE TO A FRIEND
A145 Sigma-Aldrich

1-Allyl-3,7-dimethyl-8-sulfophenylxanthine

solid

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   CDMZOKMMANFJMU-UHFFFAOYSA-N
form   solid
color   white
solubility   H2O: soluble
absorption   εmax/203 nm, methanol 28,200
  εmax/239 nm, methanol 21,900
  εmax/301 nm, methanol 18,200
Gene Information   human ... ADORA2A(135), ADORA2B(136)
rat ... Adora1(29290), Adora2a(25369)

Description

Biochem/physiol Actions

Weak A2 adenosine receptor antagonist.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?