• USA Home
  • A1549 - 4′-Acetamidophenyl 4-guanidinobenzoate hydrochloride

EMAIL THIS PAGE TO A FRIEND
A1549 Sigma

4′-Acetamidophenyl 4-guanidinobenzoate hydrochloride

  • CAS Number 79119-49-6

  • Empirical Formula (Hill Notation) C16H16N4O3 · HCl

  • Molecular Weight 348.78

  •  MDL number MFCD00078843

  •  PubChem Substance ID 24890560

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Chromogenic, Enzyme Substrates, Enzymes, Inhibitors, and Substrates More...
InChI Key   CBYYAOPEHXAUHL-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DG2800520

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?