• USA Home
  • A164 - Alaproclate hydrochloride

EMAIL THIS PAGE TO A FRIEND
A164 Sigma

Alaproclate hydrochloride

Synonym: DL-Alanine 2-(4-chlorophenyl)-1,1-dimethylethyl ester hydrochloride

  • CAS Number 60719-83-7

  • Empirical Formula (Hill Notation) C13H18ClNO2 · HCl

  • Molecular Weight 292.20

  •  MDL number MFCD00153761

  •  PubChem Substance ID 24278076

Purchase

Properties

Related Categories Biochemicals and Reagents, Biogenic Amine Transport Inhibitors, Cell Biology, Cell Signaling and Neuroscience, GPCR Modulators,
InChI Key   OPAKSOWFKIUFNP-UHFFFAOYSA-N
form   crystalline
color   white
solubility   H2O: soluble
Gene Information   human ... HTR1A(3350), HTR1B(3351), HTR1D(3352), HTR1E(3354), HTR1F(3355), HTR2A(3356), HTR2B(3357), HTR2C(3358), HTR3A(3359), HTR3B(9177), HTR3C(170572), HTR3D(200909), HTR3E(285242), HTR4(3360), HTR5A(3361), HTR5B(645694), HTR6(3362), HTR7(3363)

Description

Biochem/physiol Actions

Potent and selective serotonin uptake inhibitor.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
AY4397300

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?