• USA Home
  • A1857 - Anti-ACTR6 (381-396) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A1857 Sigma-Aldrich

Anti-ACTR6 (381-396) antibody produced in rabbit

IgG fraction of antiserum, PBS solution

Synonym: Anti-ARP6, Anti-Actin-related protein 6 homolog, Anti-CDA12, Anti-HSPC281, Anti-MSTP136, Anti-hARPX

Purchase

Properties

conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   indirect immunofluorescence: suitable
  western blot: 1:500-1:2,000
species reactivity   human
mol wt   antigen ~45 kDa
form   PBS solution
shipped in   dry ice
storage temp.   −20°C
antibody form   IgG fraction of antiserum
antibody product type   primary antibodies
UniProt accession no.   Q9GZN1
Gene Information   human ... ACTR6(64431)

Description

General description

ACTR6 (381-396) is an ARP6 actin-related homolog (yeast). Actin is an ubiquitous protein involved in the formation of filaments that are a major component of the cytoskeleton. ARP6, in vertebrate cells, is found localized to the nucleus where it has a role in maintaining chromatin organization and structure. ARP6 binds to heterochromatin protein 1 and has a role in telomere silencing in yeast.

Rabbit anti-ACTR6 (381-396) antibody binds to human ACTR6 (381-396).

Immunogen

synthetic peptide coresponding to amino acids 381-396 of human ACTR6.

Application

Rabbit anti-ACTR6 (381-396) antibody can be used for indirect immunofluorescence and western blot (1:500-1:2,000) applications.

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.2, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?