• USA Home
  • A201 - cis-4-Aminocrotonic acid

EMAIL THIS PAGE TO A FRIEND
A201 Sigma-Aldrich

cis-4-Aminocrotonic acid

solid

Synonym: 4-Amino-cis-2-butenoic acid, CACA

Purchase

Properties

form   solid
color   white
solubility   H2O: 124 mg/mL
SMILES string   [H]\C(CN)=C(/[H])C(O)=O

Description

Biochem/physiol Actions

GABAC receptor agonist

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?