• USA Home
  • A2106 - Monoclonal Anti-AGB1 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
A2106 Sigma-Aldrich

Monoclonal Anti-AGB1 antibody produced in mouse

~2 mg/mL, clone AGB1-47, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-Arabidopsis G-protein β subunit, Anti-ELK4, Anti-Erecta-like 4, Anti-GTP Binding Protein β 1, Anti-T4L20.40, Anti-T4L20_40

Purchase

Properties

Related Categories AE-AK, Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to G Proteins,
conjugate   unconjugated
clone   AGB1-47, monoclonal
biological source   mouse
application(s)   indirect ELISA: suitable
  western blot: 0.5-1 μg/mL using recombinant AGB1 protein
species reactivity   Arabidopsis sp.
mol wt   antigen ~40 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG1
antibody product type   primary antibodies
concentration   ~2 mg/mL
Gene Information   Arabidopsis thaliana ... AGB1(829597)

Description

General description

AGB1 is encoded by the gene ELK4. AGB1 is a putative β subunit heterotrimeric G-protein involved in signal transduction in animal cells.

Monoclonal anti-AGB1 recognizes Arabidopsis AGB1. The antibody does not cross react with Arabidopsis AGG1.

Immunogen

synthetic peptide corresponding to amino acids 362-377 of Arabidopsis AGB1.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Mouse monoclonal anti-AGB1 antibody can be used for western blot (0.5-1μg/mL, using recombinant AGB1 protein) and indirect ELISA assays.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?