EMAIL THIS PAGE TO A FRIEND
A2500 Sigma-Aldrich

Phenacetin

powder

Synonym: 1-Acetyl-p-phenetidin, 4′-Ethoxyacetanilide, N-(4-Ethoxyphenyl)acetamide, p-Acetophenetidide, Acetophenetidin

  • CAS Number 62-44-2

  • Linear Formula CH3CONHC6H4OC2H5

  • Molecular Weight 179.22

  •  Beilstein/REAXYS Number 1869238

  •  EC Number 200-533-0

  •  MDL number MFCD00009094

  •  PubChem Substance ID 24890660

Purchase

Properties

form   powder
mp   133-136 °C (lit.)
storage temp.   2-8°C
SMILES string   CCOc1ccc(NC(C)=O)cc1
InChI   1S/C10H13NO2/c1-3-13-10-6-4-9(5-7-10)11-8(2)12/h4-7H,3H2,1-2H3,(H,11,12)
InChI key   CPJSUEIXXCENMM-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Substrate of CYP1A2 and CYP2D6.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
RTECS 
AM4375000
Protocols & Articles

Articles

Carcinogenesis and Epigenetics

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete. The expansion of the carcinogenesi...
Vicki Caligur
BioFiles 2008, 3.5, 18.
Keywords: Acetylations, Adhesion, Alkylations, Apoptosis, Biofiles, Cancer, Carcinogens, Cell division, Cell proliferation, Cell signaling, DNA microarrays, Drug discovery, Environmental, Epigenetics, Events, Gene expression, Genetic, Hormones, Metabolism, Methylations, Microarray Analysis, Mutagens, PAGE, Recombination, Reductions, Sequences, Transcription, Type, transformation

Related Content

Carcinogen Selector - Digestive System - Stomach

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Stomach
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Digestive System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Ear Zymbal's Gland

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Mammary Gland

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Nasal Cavity

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Urinary Bladder

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Respiratory System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Urinary System - Kidney

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Urinary System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?