• USA Home
  • A3012 - Actinomycin I from Streptomyces antibioticus

EMAIL THIS PAGE TO A FRIEND
A3012 Sigma

Actinomycin I from Streptomyces antibioticus

~80% (HPLC)

  • CAS Number 18865-46-8

  • Empirical Formula (Hill Notation) C61H90N12O17

  • Molecular Weight 1263.44

  •  MDL number MFCD00067539

Purchase

Properties

assay   ~80% (HPLC)
Mode of action   DNA synthesis | interferes
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
AU1599333

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?