• USA Home
  • A3362 - Cisatracurium besylate

EMAIL THIS PAGE TO A FRIEND
A3362 Sigma-Aldrich

Cisatracurium besylate

  • CAS Number 96946-42-8

  • Empirical Formula (Hill Notation) C53H72N2O12·2C6H5O3S

  • Molecular Weight 1243.48

  •  MDL number MFCD00871018

  •  PubChem Substance ID 329770719

Purchase

Properties

Related Categories Agonists, Cell Biology, Cell Signaling and Neuroscience, Cholinergics, Ion Channels,
form   (Powder)
color   white
mp   85-90 °C (lit.)
solubility   H2O: 1 mg/mL
Mode of action   cell membrane | interferes
storage temp.   −20°C
SMILES string   [O-]S(=O)(=O)c1ccccc1.[O-]S(=O)(=O)c2ccccc2.COc3ccc(C[C@@H]4c5cc(OC)c(OC)cc5CC[N@+]4(C)CCC(=O)OCCCCCOC(=O)CC[N@@+]6(C)CCc7cc(OC)c(OC)cc7[C@H]6Cc8ccc(OC)c(OC)c8)cc3OC
InChI   1S/C53H72N2O12.2C6H6O3S/c1-54(22-18-38-32-48(62-7)50(64-9)34-40(38)42(54)28-36-14-16-44(58-3)46(30-36)60-5)24-20-52(56)66-26-12-11-13-27-67-53(57)21-25-55(2)23-19-39-33-49(63-8)51(65-10)35-41(39)43(55)29-37-15-17-45(59-4)47(31-37)61-6;2*7-10(8,9)6-4-2-1-3-5-6/h14-17,30-35,42-43H,11-13,18-29H2,1-10H3;2*1-5H,(H,7,8,9)/q+2;;/p-2/t42-,43-,54-,55-;;/m1../s1
InChI key   XXZSQOVSEBAPGS-DONVQRBFSA-L
Gene Information   human ... CHRNA1(1134), CHRNB1(1140), CHRND(1144), CHRNE(1145), CHRNG(1146)

Description

Packaging

10 mg in glass bottle

100 mg in poly bottle

Biochem/physiol Actions

Cisatracurium besylate is a potent, nondepolarizing neuromuscular blocking agent. It acts on cholinergic receptors to block neuromuscular transmission, which can be antagonized by acetylcholinesterase inhibitors. Cisatracurium besylate is one of the ten stereoisomers of astracurium that does not cause histamine release.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?