• USA Home
  • A4404 - Anti-Arachis hypogaea Lectin antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A4404 Sigma

Anti-Arachis hypogaea Lectin antibody produced in rabbit

fractionated antiserum, lyophilized powder

Synonym: Anti-PNA, Anti-Peanut Lectin

Purchase

Properties

Related Categories AR-AR, Alphabetical Index, Antibodies, Cell Biology, Cell Signaling and Neuroscience,
biological source   rabbit
antibody form   fractionated antiserum
clone   polyclonal
form   lyophilized powder
packaging   vial of ~2 mL Lyophilized antiserum
application(s)   indirect ELISA: 1:80,000
conjugate   unconjugated
storage temp.   2-8°C

Description

Immunogen

Peanut lectin (PNA)

Reconstitution

Reconstitute with 2 mL deionized water.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2.

Application

Mouse myotube cultures were ran on SDS page gels and membranes treated with PNA for 24hrs the probed with anti-PNA anitbody at a 1:10 dilution.

Rabbit Anti-Arachis hypogaea Lectin antibody can be used for Ouchterlony double diffusion (1:8) assays.

General description

Arachis hypogaea Lectin or peanut lectin (PNA) is known to function as a marker for intercalated cells in rats. PNA also binds to glycoproteins present in human epidermal keratinocytes. Thus, anti-PNA antibodies are often used for protein localization and immunoprecipitation assays of PNA-binding cells or proteins. Rabbit Anti-Arachis hypogaea Lectin antibody binds to Arachis hypogaea lectin.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?