• USA Home
  • A5097 - DABCYL-[Asn670, Leu671]-Amyloid β/A4 Precursor Protein 770 Fragment 669-674-EDANS ammonium salt

EMAIL THIS PAGE TO A FRIEND
A5097 Sigma

DABCYL-[Asn670, Leu671]-Amyloid β/A4 Precursor Protein 770 Fragment 669-674-EDANS ammonium salt

≥95% (HPLC), solid

  • Empirical Formula (Hill Notation) C54H70N12O15S

  • Molecular Weight 1159.27

  •  MDL number MFCD03093392

  •  PubChem Substance ID 329770867

Purchase

Description

Amino Acid Sequence

DABCYL-Val-Asn-Leu-Asp-Ala-Glu-EDANS

Biochem/physiol Actions

β-Secretase substrate.

Price and Availability


Making Headway

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?