• USA Home
  • A5300 - 5α-Androstane-3α,17β-diol 17β-D-glucuronide sodium salt

EMAIL THIS PAGE TO A FRIEND
A5300 Sigma-Aldrich

5α-Androstane-3α,17β-diol 17β-D-glucuronide sodium salt

Synonym: 3α,17β-Dihydroxy-5α-androstane 17β-D-glucuronide

Purchase

Properties

drug control   USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?