EMAIL THIS PAGE TO A FRIEND
A5639 Sigma

Allyl sulfide

storage temp.: 2-8°C

Synonym: 3,3′-Thiobis-1-propene, Diallyl sulfide, NSC 20947

  • CAS Number 592-88-1

  • Linear Formula (CH2=CHCH2)2S

  • Molecular Weight 114.21

  •  Beilstein Registry Number 1736016

  •  EC Number 209-775-1

  •  MDL number MFCD00008658

Purchase

Properties

vapor density   3.9 (vs air)
vapor pressure   7 mmHg ( 20 °C)
InChI Key   UBJVUCKUDDKUJF-UHFFFAOYSA-N
refractive index   n20/D 1.490(lit.)
bp   138 °C(lit.)
mp   −83 °C (lit.)
density   0.887 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Biochem/physiol Actions

Organosulfur compound from garlic that inhibits chemically-induced carcinogenesis in experimental animals. Competitive inhibitor of Cytochrome P450 2E1 (CYP2E1) that, in turn, blocks the activation of several chemcal carcinogens.

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
2
RTECS 
BC4900000
Flash Point(F) 
114.8 °F
Flash Point(C) 
46 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

A35801 Allyl sulfide, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?