• USA Home
  • A5832 - 2-Methyl-5-nitroaniline

EMAIL THIS PAGE TO A FRIEND
A5832 Sigma-Aldrich

2-Methyl-5-nitroaniline

≥95% (GC)

Synonym: 2-Amino-4-nitrotoluene, 5-Nitro-o-toluidine

  • CAS Number 99-55-8

  • Linear Formula CH3C6H3(NO2)NH2

  • Molecular Weight 152.15

  •  Beilstein/REAXYS Number 879021

  •  EC Number 202-765-8

  •  MDL number MFCD00007741

Purchase

Properties

assay   ≥95% (GC)
mp   103-106 °C (lit.)
storage temp.   2-8°C
SMILES string   Cc1ccc(cc1N)[N+]([O-])=O
InChI   1S/C7H8N2O2/c1-5-2-3-6(9(10)11)4-7(5)8/h2-4H,8H2,1H3
InChI key   DSBIJCMXAIKKKI-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2660 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

402729 2-Methyl-5-nitroaniline, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?