• USA Home
  • A6277 - [Val4]-Angiotensin III acetate salt hydrate

EMAIL THIS PAGE TO A FRIEND
A6277 Sigma-Aldrich

[Val4]-Angiotensin III acetate salt hydrate

≥97% (HPLC)

  • Empirical Formula (Hill Notation) C45H64N12O9 · xC2H4O2 · yH2O

  • Molecular Weight 917.06 (anhydrous free base basis)

  •  MDL number MFCD11046001

  •  PubChem Substance ID 329770888

Purchase

Properties

Related Categories Angiotensins, Cell Biology, Cell Signaling and Neuroscience, Neuropeptides, Peptides and Proteins,
assay   ≥97% (HPLC)
storage temp.   −20°C
SMILES string   O.CC(O)=O.CC(C)[C@H](NC(=O)[C@@H](N)CCCNC(N)=N)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N3CCC[C@H]3C(=O)N[C@@H](Cc4ccccc4)C(O)=O
InChI   1S/C45H64N12O9.C2H4O2.H2O/c1-25(2)36(55-38(59)31(46)12-8-18-50-45(47)48)41(62)52-32(20-28-14-16-30(58)17-15-28)39(60)56-37(26(3)4)42(63)53-33(22-29-23-49-24-51-29)43(64)57-19-9-13-35(57)40(61)54-34(44(65)66)21-27-10-6-5-7-11-27;1-2(3)4;/h5-7,10-11,14-17,23-26,31-37,58H,8-9,12-13,18-22,46H2,1-4H3,(H,49,51)(H,52,62)(H,53,63)(H,54,61)(H,55,59)(H,56,60)(H,65,66)(H4,47,48,50);1H3,(H,3,4);1H2/t31-,32-,33-,34-,35-,36-,37-;;/m0../s1
InChI key   QVJGCLYZCFQPQT-PGCZNNRSSA-N

Description

Amino Acid Sequence

Arg-Val-Tyr-Val-His-Pro-Phe

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?