• USA Home
  • A6648 - Adenosine Deaminase from bovine spleen

EMAIL THIS PAGE TO A FRIEND
A6648 Sigma

Adenosine Deaminase from bovine spleen

Type V, buffered aqueous glycerol solution, 60-130 units/mg protein

Synonym: Adenosine aminohydrolase

Purchase

Properties

Related Categories 3.5.x.x Acting on C-N other than peptides, 3.x.x.x Hydrolases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
type   Type V
form   buffered aqueous glycerol solution
cation traces   NH4+: <25 μg/mg protein
foreign activity   alkaline phosphatase, essentially free
storage temp.   2-8°C
Gene Information   cow ... ADA(280712)

Description

Analysis Note

Protein determined by biuret.

Unit Definition

One unit will deaminate 1.0 μmole of adenosine to inosine per min at pH 7.5 at 25 °C.

Physical form

Solution in 50% glycerol, 50 mM potassium phosphate, pH 6.0

General description

Molecular weight: 32.5-33 kDa

pI: 4.85

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

A5043 Adenosine Deaminase from bovine spleen, Type X, buffered aqueous glycerol solution, ≥130 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?