• USA Home
  • A7414 - Anti-Bovine IgG (whole molecule)−Peroxidase antibody produced in rabbit

A7414 Sigma

Anti-Bovine IgG (whole molecule)−Peroxidase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution



Related Categories Alphabetical Index, Antibodies, BL-BZ, Bovine Secondary Antibodies and Conjugates, Peroxidase Labeled Antibodies,
biological source   rabbit
antibody form   affinity isolated antibody
clone   polyclonal
form   buffered aqueous solution
application(s)   direct ELISA: 1:10,000
conjugate   peroxidase conjugate
shipped in   dry ice
storage temp.   −20°C


Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in "Immunofluorescence and Related Staining Techniques," Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.01% thimerosal.

Price and Availability

Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany 
Protocols & Articles


Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers


Related Products

comparable product

Product #


Add to Cart

A5295 Anti-Bovine IgG (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?