EMAIL THIS PAGE TO A FRIEND
A8259 Sigma-Aldrich

α-Asarone

Synonym: trans-1,2,4-Trimethoxy-5-(1-propenyl)benzene, trans-1-Propenyl-2,4,5-trimethoxybenzene, trans-Asarone, Isoasaron

  • CAS Number 2883-98-9

  • Linear Formula (CH3O)3C6H2CH=CHCH3

  • Molecular Weight 208.25

  •  Beilstein/REAXYS Number 1910606

  •  EC Number 220-743-6

  •  MDL number MFCD00064457

Purchase

Properties

bp   296 °C (lit.)
mp   57-61 °C (lit.)
storage temp.   −20°C
SMILES string   COc1cc(OC)c(\C=C\C)cc1OC
InChI   1S/C12H16O3/c1-5-6-9-7-11(14-3)12(15-4)8-10(9)13-2/h5-8H,1-4H3/b6-5+
InChI key   RKFAZBXYICVSKP-AATRIKPKSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DC2975000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?