EMAIL THIS PAGE TO A FRIEND
A9164 Sigma-Aldrich

Ammonium persulfate

for molecular biology, powder, ≥98%

Synonym: AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER

Purchase

Properties

Related Categories Automated Sequencing, Chemical Synthesis, Gel Solutions and Reagents, Molecular Biology, Oxidizing Agents,
grade   for molecular biology
vapor density   7.9 (vs air)
assay   ≥98%
form   powder
solubility   H2O: soluble
anion traces   chloride (Cl-): <10 ppm
cation traces   Fe: <10 ppm
  heavy metals (as Pb): <50 ppm
SMILES string   N.N.OS(=O)(=O)OOS(O)(=O)=O
InChI   1S/2H3N.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h2*1H3;(H,1,2,3)(H,4,5,6)
InChI key   ROOXNKNUYICQNP-UHFFFAOYSA-N

Description

Application

Catalyst for acrylamide gel polymerization.

Reconstitution

10% stock solution may be stored at 4 °C for several weeks.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1444 5.1 / PGIII
WGK Germany 
1
RTECS 
SE0350000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?