EMAIL THIS PAGE TO A FRIEND
A9236 Sigma-Aldrich

AQ4

≥98% (HPLC)

Synonym: 1,4-Bis[[2-(dimethylamino)ethyl]amino]-5,8-dihydroxy-9,10-anthracenedione, 1,4-Bis[[2-(dimethylamino)ethyl]amino]-5,8-dihydroxyanthracene-9,10-dione

Purchase

Properties

Related Categories AN-AZ, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology
InChI Key   DBAMBJQJKDHEQX-UHFFFAOYSA-N
assay   ≥98% (HPLC)
form   powder
storage condition   desiccated
color   dark blue
solubility   DMSO: ≥10 mg/mL
storage temp.   2-8°C

Description

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

AQ4 is a chemically stable cytotoxic agent which acts as a topoisomerase II inhibitor and DNA intercalator. The compound is a potent cytotoxic agent in many human tumor lines, including those derived from human pancreatic adenocarcinomas. The compound N-oxide is a hypoxia activated prodrug of topoisomerase-targeting.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?