• USA Home
  • A9273 - Ala-Ala-Val-Ala p-nitroanilide

EMAIL THIS PAGE TO A FRIEND
A9273 Sigma-Aldrich

Ala-Ala-Val-Ala p-nitroanilide

protease substrate

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, Chymotrypsin, alpha, Enzyme Substrates,
storage temp.   2-8°C
SMILES string   CC(C)C(NC(=O)C(C)NC(=O)C(C)N)C(=O)NC(C)C(=O)Nc1ccc(cc1)N(=O)=O
InChI   1S/C20H30N6O6/c1-10(2)16(25-19(29)13(5)22-17(27)11(3)21)20(30)23-12(4)18(28)24-14-6-8-15(9-7-14)26(31)32/h6-13,16H,21H2,1-5H3,(H,22,27)(H,23,30)(H,24,28)(H,25,29)
InChI key   VEDBEYLQKXTZOG-UHFFFAOYSA-N
Gene Information   rat ... Npy(24604)

Description

Substrates

Substrate for chymotrypsin and tripeptidyl peptidase I.

Safety & Documentation

Safety Information

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?