EMAIL THIS PAGE TO A FRIEND
A9812 Supelco

6-Aminocaproic acid

Sigma Reference Standard

Synonym: ε-Aminocaproic acid, 6-Aminohexanoic acid, EACA

  • CAS Number 60-32-2

  • Linear Formula H2N(CH2)5CO2H

  • Molecular Weight 131.17

  •  Beilstein/REAXYS Number 906872

  •  EC Number 200-469-3

  •  MDL number MFCD00008238

Purchase

Properties

grade   Sigma Reference Standard
packaging   vial of 200 mg
mp   207-209 °C (dec.) (lit.)
SMILES string   NCCCCCC(O)=O
InChI   1S/C6H13NO2/c7-5-3-1-2-4-6(8)9/h1-5,7H2,(H,8,9)
InChI key   SLXKOJJOQWFEFD-UHFFFAOYSA-N
Gene Information   human ... PLG(5340)

Description

Biochem/physiol Actions

Lysine analog. Promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis. Reported to inhibit plasminogen binding to activated platelets. An early report indicated that it inhibits the activation of the first component of the complement system. Binds and inactivates Carboxypeptidase B.

Packaging

Supplied in amber screw-cap vials

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
2
RTECS 
MO6300000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

A7824 6-Aminocaproic acid, BioUltra, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?