• USA Home
  • APREST71281 - PrEST Antigen C1orf116

EMAIL THIS PAGE TO A FRIEND
APREST71281 Sigma-Aldrich

PrEST Antigen C1orf116

Prestige Antigens Powered by Atlas Antibodies, buffered aqueous solution

Synonym: FLJ36507, MGC2742, MGC4309, SARG

Purchase

Properties

Related Categories Antibodies, Prestige Antigens, Prestige Antigens C - D More...
Quality Level   100
recombinant   expressed in E. coli
assay   >80% (SDS-PAGE)
form   buffered aqueous solution
mol wt   predicted mol wt 34 kDa
purified by   immobilized metal affinity chromatography (IMAC)
concentration   ≥0.5 mg/mL
immunogen sequence   ANSALTPPKPESGLTLQESNTPGLRQMNFKSNTLERSGVGLSSYLSTEKDASPKTSTSLGKGSFLDKISPSVLRNSRPRPASLGTGKDFAGIQVGKLADLEQEQSSKRLSYQGQSRDKLPRPPCVSVKISPKGVPNEHRREALKKLGLL
Ensembl | human accession no.   ENSG00000182795
UniProt accession no.   Q9BW04
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... C1orf116(79098)

Description

General description

Recombinant protein fragment of Human C1ORF116 with N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag.

Application

Suitable as a blocking agent using corresponding antibodies.

Linkage

Corresponding Antibody HPA011888.

Physical form

Solution in 1 M urea-PBS, pH 7.4

Preparation Note

The protein solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user.

Legal Information

Prestige Antigens is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?