• USA Home
  • APREST89801 - PrEST Antigen PRDM8

EMAIL THIS PAGE TO A FRIEND
APREST89801 Sigma-Aldrich

PrEST Antigen PRDM8

Prestige Antigens Powered by Atlas Antibodies, buffered aqueous solution

Purchase

Properties

Related Categories Antibodies, Prestige Antigens, Prestige Antigens O - P More...
recombinant   expressed in E. coli
assay   >80% (SDS-PAGE)
form   buffered aqueous solution
mol wt   predicted mol wt 25 kDa
purified by   immobilized metal affinity chromatography (IMAC)
concentration   ≥0.5 mg/mL
immunogen sequence   RCPKRLHSADISPQDEQGGGVGTKDHGGGGGGGKDQQQQQQEAPLGPGPKFCKAGPLHHYPSPSPESSNPS
Ensembl | human accession no.   ENSG00000152784
UniProt accession no.   Q9NQV8
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... PRDM8(56978)

Description

Application

Blocking agent and positive assay control using corresponding antibodies.

Linkage

Corresponding Antibody HPA057253.

Preparation Note

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.

Physical form

Solution in phosphate-buffered saline and 1M Urea, pH 7.4

General description

Recombinant protein fragment of Human PRDM8 with N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag.    

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary and Secondary Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay needs. Use the Antibody...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?