• USA Home
  • AQ0045 - AQUA Peptide LQEEIDAVLPN[K]

EMAIL THIS PAGE TO A FRIEND
AQ0045 Supelco

AQUA Peptide LQEEIDAVLPN[K]

  • Empirical Formula (Hill Notation) C5413C6H101N1315N2O21

  • Molecular Weight 1376.47

  •  NACRES NA.26

Purchase

Properties

Related Categories AQUA Peptides, Mass Spectrometry, Molecular Biology, Proteomics
mol wt   monoisotopic mol wt 1375.74 Da
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... CYP3A4(1576), CYP3A43(64816)

Description

Application

Protein-AQUA is a powerful, enabling technology that facilitates focused, quantitative studies of not only specific protein expression, but specific amino acid modification as well, using stable-isotope–labeled peptides and mass spectrometry.

Legal Information

This method was developed by Dr. Steve Gygi and colleagues at Harvard Medical School. Limited use of this method is permitted under a licensing arrangement with Harvard Medical School.

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company.  The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration.  The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party.  Information about licenses for excluded uses is available from:  E. I. du Pont de Nemours and Company;  Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), licensing@dupont.com.

AQUA is a trademark of Harvard Medical School

Protein AQUA is a trademark of Harvard Medical School

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Stable Isotope-labeled MS Internal Standards

What happens when you combine state-of-the-art technology with scientific prowess? Full-length protein standards with the highest purity and isotopic incorporation in the industry. Stable isotope-lab...
Keywords: Atomic absorption spectroscopy, Buffers, Digestions, Mass spectrometry, PAGE, Phosphorylations, Proteomics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?