• USA Home
  • AQ0341 - AQUA Peptide MCAQQLGDDCTEAAICDPH[R]

EMAIL THIS PAGE TO A FRIEND
AQ0341 Sigma

AQUA Peptide MCAQQLGDDCTEAAICDPH[R]

Purchase

Properties

mol wt   monoisotopic mol wt 2185.889 Da
shipped in   wet ice
storage temp.   −20°C

Description

Application

Protein-AQUA is a powerful, enabling technology that facilitates focused, quantitative studies of not only specific protein expression, but specific amino acid modification as well, using stable-isotope-labeled peptides and mass spectrometry.

Legal Information

This method was developed by Dr. Steve Gygi and colleagues at Harvard Medical School. Limited use of this method is permitted under a licensing arrangement with Harvard Medical School.

AQUA is a trademark of Harvard Medical School

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?