• USA Home
  • AV48141 - Anti-PGK1 (AB2) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
AV48141 Sigma-Aldrich

Anti-PGK1 (AB2) antibody produced in rabbit

IgG fraction of antiserum, lyophilized powder

Synonym: Anti-Phosphoglycerate kinase 1, Anti-MGC117307, Anti-MGC142128, Anti-MGC8947, Anti-MIG10, Anti-PGKA

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PD-PG, Primary Antibodies
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunohistochemistry: suitable
  western blot: suitable
species reactivity   rabbit, canine, horse, human, rat, mouse, pig, bovine, zebrafish
mol wt   44 kDa
form   lyophilized powder
storage temp.   −20°C
antibody form   IgG fraction of antiserum
antibody product type   primary antibodies
immunogen sequence   TADKFDENAKTGQATVASGIPAGWMGLDCGPESSKKYAEAVTRAKQIVWN
NCBI accession no.   NP_000282
UniProt accession no.   P00558
Gene Information   human ... PGK1(5230)

Description

General description

Phosphoglycerate kinase 1 (PGK1) is an enzyme that catalyzes the conversion of 1,3-diphosphoglycerate to 3-phosphoglycerate during glycolysis. PGK1 expression has been correlated to CXCR4 expression, survival and bone marrow metastasis in neuroblastoma patients.
Rabbit Anti-PGK1 antibody recognizes human, rat, bovine, pig, zebrafish, canine, and mouse PGK1.

Immunogen

synthetic peptide corresponding to a region of human PGK1 with an internal ID of S22609

Application

Rabbit Anti-PGK1 antibody is suitable for western blot applications at a concentration of 1.25μg/ml and for IHC at 4-8μg/ml using paraffin-embedded tissues.

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?