• USA Home
  • AV48226 - Anti-VIM (AB2) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
AV48226 Sigma-Aldrich

Anti-VIM (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym: Anti-FLJ36605, Anti-Vimentin

  •  NACRES NA.41

Purchase

Properties

conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunohistochemistry: suitable
  western blot: suitable
species reactivity   dog, sheep, guinea pig, rabbit, rat, horse, bovine, human, mouse
mol wt   54 kDa
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
Quality Level   100
antibody product type   primary antibodies
concentration   0.5 mg - 1 mg/mL
NCBI accession no.   NP_003371
UniProt accession no.   P08670
Gene Information   human ... VIM(7431)

Description

General description

Vimentin (VIM) codes for an intermediate filament protein that modulates cell shape, cytoplasm integrity and cytoskeletal associations. It also regulates immune response, cell signaling and cellular transport. VIM has been analyzed as a molecular target for the treatment of cancer and hepatitis B virus related hepatocellular carcinoma. Vimentin is known to induce malignant transformation in metastatic melanoma.
Rabbit Anti-VIM antibody recognizes bovine, pig, chicken, zebrafish, human, mouse, and rat VIM.

Immunogen

Synthetic peptide directed towards the C terminal region of human VIM

Application

Rabbit Anti-VIM antibody has been used for western blot applications.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Biochem/physiol Actions

Along with the microfilaments (actins) and microtubules (tubulins), the intermediate filaments represent a third class of well-characterized cytoskeletal elements. The subunits display a tissue-specific pattern of expression. Desmin is the subunit specific for muscle and vimentin the subunit specific for mesenchymal tissue.Along with the microfilaments (actins) and microtubules (tubulins), the intermediate filaments represent a third class of well-characterized cytoskeletal elements. The subunits display a tissue-specific pattern of expression. Desmin (MIM 125660) is the subunit specific for muscle and vimentin the subunit specific for mesenchymal tissue.[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: SSLNLRETNLDSLPLVDTHSKRTLLIKTVETRDGQVINETSQHHDDLE

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?