• USA Home
  • AV50601 - Anti-PYGO1 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
AV50601 Sigma-Aldrich

Anti-PYGO1 antibody produced in rabbit

affinity isolated antibody

Synonym: Anti-DKFZp547G0910, Anti-Pygopus homolog 1 (Drosophila)

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PT-PY, Primary Antibodies
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: suitable
species reactivity   human, horse, bovine, dog, rabbit, guinea pig, rat, mouse
mol wt   45 kDa
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
Quality Level   100
antibody product type   primary antibodies
concentration   0.5 mg - 1 mg/mL
NCBI accession no.   NP_056432
UniProt accession no.   Q9Y3Y4
Gene Information   human ... PYGO1(26108)

Description

General description

PYGO1 codes for pygopus family PHD finger 1. Studies in mice have revealed that Pygo1 is needed for branching morphogenesis during renal development. It is also involved in Wnt signaling pathways.
Rabbit Anti-PYGO1 antibody recognizes human, mouse, zebrafish, and chicken PYGO1.

Immunogen

Synthetic peptide directed towards the N terminal region of human PYGO1

Application

Rabbit Anti-PYGO1 antibody is suitable for western blot applications at a concentration of 1μg/ml.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Biochem/physiol Actions

PYGO1 contains 1 PHD-type zinc finger. PYGO1 is involved in signal transduction through the Wnt pathway.

Sequence

Synthetic peptide located within the following region: GGPGVQLGSPDKKKRKANTQGPSFPPLSEYAPPPNPNSDHLVAANPFDDN

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?