• USA Home
  • B0885 - Nα-Benzoyl-L-arginine 4-methoxy-β-naphthylamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
B0885 Sigma-Aldrich

Nα-Benzoyl-L-arginine 4-methoxy-β-naphthylamide hydrochloride

  • CAS Number 100900-33-2

  • Empirical Formula (Hill Notation) C24H27N5O3 · HCl

  • Molecular Weight 469.96

  •  MDL number MFCD00058038

  •  PubChem Substance ID 24891536

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?