EMAIL THIS PAGE TO A FRIEND
B1000 Sigma-Aldrich

Bathocuproine

Synonym: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, BCP

  • CAS Number 4733-39-5

  • Empirical Formula (Hill Notation) C26H20N2

  • Molecular Weight 360.45

  •  Beilstein/REAXYS Number 306714

  •  EC Number 225-240-5

  •  MDL number MFCD00004972

Purchase

Properties

mp   279-283 °C (lit.)
SMILES string   Cc1cc(-c2ccccc2)c3ccc4c(cc(C)nc4c3n1)-c5ccccc5
InChI   1S/C26H20N2/c1-17-15-23(19-9-5-3-6-10-19)21-13-14-22-24(20-11-7-4-8-12-20)16-18(2)28-26(22)25(21)27-17/h3-16H,1-2H3
InChI key   STTGYIUESPWXOW-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

140910 Bathocuproine, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?