• USA Home
  • B1026 - 5-Bromo-4-chloro-3-indolyl phosphate disodium salt

EMAIL THIS PAGE TO A FRIEND
B1026 Sigma

5-Bromo-4-chloro-3-indolyl phosphate disodium salt

for molecular biology, ≥98%, powder

Synonym: 5-Bromo-4-chloro-3-indolyl phosphate disodium salt, BCIP®, X-phosphate disodium salt

Purchase

Properties

grade   for molecular biology
InChI Key   OAZUOCJOEUNDEK-UHFFFAOYSA-L
assay   ≥98%
form   powder
solubility   H2O: 20 mg/mL
  DMF: insoluble
foreign activity   Protease, none detected
storage temp.   −20°C

Description

Substrates

Histochemical substrate for alkaline phosphatase.

Application

For the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.

BCIP is provided in two salt forms: the disodium salt which is soluble in water and the p-toluidine form which is soluble in dimethylformamide. These salt forms may be used to prepare a stock solution that in combination with NBT and a reaction buffer, form a substrate solution for alkaline phosphatase. This substrate system, when incubated with alkaline phosphatase, produces an insoluble NBT diformazan product that is easily observable with its purple color. For added convenience, a mixture of BCIP and NBT is provided in an easily dissolvable tablet form or as a ready-to-use liquid.

Legal Information

BCIP is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?