• USA Home
  • B1559 - Anti-Bmf, C-Terminal antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
B1559 Sigma-Aldrich

Anti-Bmf, C-Terminal antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym: Anti-Bcl-2-modifying factor

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Bcl-2 Family Proteins,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   microarray: suitable
  western blot: 2.5 μg/mL
species reactivity   mouse, human
mol wt   antigen ~25 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
concentration   ~1 mg/mL
UniProt accession no.   Q96LC9
Gene Information   human ... BMF(90427)
mouse ... Bmf(171543)

Description

Immunogen

synthetic peptide corresponding to amino acids 171-184 of human Bmf.

Application

Polyclonal anti-Bmf, N-Terminal antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/mL. It is also suitable for microarray analysis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Biochem/physiol Actions

Bmf(Bcl-2-modifying factor) gene encodes a BH3-only protein that belongs to BCL2 protein family and is localized to chromosome 15 at 15q14. Isoform 1 of this gene is primarily expressed in B-lymphoid cells whereas Isoform 2 and isoform 3 are chiefly expressed in B-CLL and normal B-cells. The BH3 domain in Bmf is essential for binding to Bcl-2 proteins as well as for triggering apoptosis. In healthy cells, Bmf interacts with the dynein light chain 2 (DLC2) component of the myosin V motors and is sequestered by actin cytoskeleton in the cell. Disruption of the actin cytoskeleton, either by depolymerization of actin filaments or by detachment of cells from the extracellular matrix, initiates activation and release of Bmf, initiating the downstream apoptotic program.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?