EMAIL THIS PAGE TO A FRIEND
B1642 Sigma-Aldrich

Boc-Arg(Mtr)-OH

Synonym: Nα-Boc-Nω-(4-methoxy-2,3,6-trimethylbenzenesulfonyl)-L-arginine, Nα-Boc-Nω-MTR-L-arginine

  • CAS Number 102185-38-6

  • Empirical Formula (Hill Notation) C21H34N4O7S

  • Molecular Weight 486.58

  •  Beilstein/REAXYS Number 7060101

  •  MDL number MFCD00043097

Purchase

Properties

assay   >99% (TLC)
storage temp.   −20°C
SMILES string   COc1cc(C)c(c(C)c1C)S(=O)(=O)NC(=N)NCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?