• USA Home
  • B21201 - EnPresso® B Defined Nitrogen-free

EMAIL THIS PAGE TO A FRIEND
B21201 Sigma

EnPresso® B Defined Nitrogen-free

Growth system for expressing protein in bacteria

Purchase

Properties

Related Categories Bacterial Culture Media, Cell Biology, EnPresso Growth Systems for microbial culture, Life Science Research, Microbial Media for Molecular Biology and Protein Expression,
sterility   sterile; γ-irradiated
form   tablet
pH   6.8
shipped in   ambient
storage temp.   room temp

Description

Biochem/physiol Actions

EnPresso® B Defined Nitrogen-free is a pre-sterilized, chemically-defined growth system designed to increase the yield of 15N-labelled proteins.

EnPresso® B growth systems provide optimal conditions for growth, metabolism and protein expression in microbial cultures. Protein yields are increased by enabling cultures to reach far higher cell densities than those achieved using conventional media. By controlling growth rate and metabolism, a greater proportion of expressed protein can be correctly folded to improve solubility, minimize the risk of inclusion body formation, and ensure functionality of the final product.

EnPresso® B growth systems maintain pH, provide adequate minerals, vitamins and trace elements to support growth, and use proprietary EnBase technology to ensure a constant, slow release of glucose from a polysaccharide substrate.

See all available products from EnPresso B Growth Systems.

Physical form

EnPresso® B is supplied in a kit providing sufficient reagents for 20 separate 50 ml cultures. Included in the kit:
40 tablets in 20 blue bags
1 bottle (5 ml) Reagent A

Legal Information

EnBase is a trademark of BioSilta Oy

EnPresso is a registered trademark of BioSilta Oy

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

EnBase Technology - how it works (with webinar)

EnBase technology is a key element within every EnPresso® growth system, offering a novel approach to control the feeding, and thereby growth, of microbial cultures. A glucose-releasing agent breaks ...
Keywords: Deaminations, Metabolism, Metabolites, Respiration

EnPresso FAQs

A. EnPresso B growth system ready for use. Note small crystals of magnesium salts are visible – these do not affect performance.
Keywords: Anaerobic, Antibiotics, Cell disruption, Degradations, Gene expression, Indicators, Metabolism, Nutrition, Protein extraction, Purification, Reductions, Respiratory, Tagged proteins

EnPresso® B Defined Nitrogen-free (E. coli protein data)

Poor growth, low levels of expression and insufficient labeled protein are the most frequent challenges faced when using minimal medium to prepare proteins for structural characterization by NMR anal...
Keywords: Gene expression, Nuclear magnetic resonance spectroscopy, Pharmaceutical, Purification

EnPresso® Growth Systems: Revolutionizing the Role of Microbial Cell Culture

Growing cells to express proteins may seem just a small step on the way to achieving your research goals, but…
Keywords: Cell culture, Metabolites

Improvements in Yield of 15N labelled proteins

Nuclear Magnetic Resonance (NMR) spectroscopy is an important technique for structure–based drug design. NMR can both characterise protein molecules and be used as a screening tool for the binding of...
Julia Smith, Ben Davies
Vernalis (R&D) Ltd, Granta Park, Cambridge, CB21 6GB
Keywords: Antibiotics, Ligands, Nuclear magnetic resonance spectroscopy, Purification, Spectroscopy, Vitamins, transformation

Introduction to Microbial Media

Microbiological studies require the availability of different species of bacteria, yeast or viruses in the laboratory. Large scale processes such as fermentation, protein and vaccine production requi...
Keywords: Antibiotics, Cellular processes, Cloning, Culture media, Fermentation, Gene expression, Growth factors, Indicators, Molecular biology, Nutrition, Sterilizations, Vitamins, transformation

Production of 15N-13C dual-labelled proteins

One challenge in NMR is the need to produce higher yields of functionally labelled recombinant protein, and then isotopically label these for rapid spectral analysis in structural studies.
Dr. Alan Robertson and Dr. Ben Davis,
Vernalis, UK Ltd.
Keywords: Drug discovery, Ligands, Nuclear magnetic resonance spectroscopy, Purification, Spectroscopy

Protein Expression Systems

The development of genetic engineering and cloning has opened many possibilities of expression and isolation of heterologous proteins for research purposes. Considerable advances in technology have e...
Keywords: Amplification, Antibiotics, Cell disruption, Cloning, Culture media, Gene expression, Genetic, Genetics, Glycosylations, Molecular biology, Purification, Pyrogens, Transfection, transformation

Protocols

EnPresso® B Defined Nitrogen-free

EnPresso B Defined Nitrogen-free is a pre-sterilized, chemically-defined growth system designed to increase the yield of 15N-labelled proteins. EnPresso growth systems provide optimal conditions for ...
Keywords: Antibiotics, Diagnostic, Filtration, Gene expression, Metabolism, Vitamins

Related Content

Synthetic Biology Resources

Sigma® Life Science is working to use our stregnths to build the field of Synthetic Biology. Below is a collection of links to our favorite resources. Sigma also offers thousands of products for your...
Keywords: Cell culture, Cloning, Gene expression, Genetic, Genetics, Metabolic Pathways, Metabolites, Molecular biology, Molecular biology techniques, Photosynthesis, Poisons, Polymerase chain reaction - quantitative, Transcription, Transfection

Webinar: High Protein Yields in Shaken Cultures by EnPresso® Growth System

EnPresso, a proprietary growth system from BioSilta Ltd, applies slow enzymatic glucose release to control the growth and protein synthesis rate in bacteria and yeasts. This high-cell-density growth ...
Keywords: Gene expression, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

A8311 Antifoam 204, aqueous emulsion for bacterial and mammalian systems
299251 Ammonium-15N chloride, ≥98 atom % 15N, ≥99% (CP)
I1284 Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
L9234 LB Broth (Lennox), Liquid microbial growth medium

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?


Milli-Q® Water Purification Solutions