• USA Home
  • B2390 - (±)-Brompheniramine maleate salt

EMAIL THIS PAGE TO A FRIEND
B2390 Sigma

(±)-Brompheniramine maleate salt

Purchase

Properties

InChI Key   SRGKFVAASLQVBO-BTJKTKAUSA-N
Gene Information   human ... HRH1(3269)

Description

Biochem/physiol Actions

Antihistamine; H1 histamine receptor antagonist.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
US4025000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?